Would you like to log in again?
SR 450 Series Single Stage Heavy Duty Regulators, Victor
Supplier: Victor
341-0781-0527
300008-239EA 595.09 USD InStock
300008-239
SR 450 Series Single Stage Heavy Duty Regulators, Victor
Welding Supplies
  • Medium to high gas service capacity
  • Single stage design
  • Stem type seat mechanism
  • Delrin® cap bushing for smooth adjustments
  • External self reseating relief valve not designed to protect downstream apparatus
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty