Would you like to log in again?
Cylinder Holders, 4B's Bracket, ORS Nasco
Supplier: ORS Nasco
344-TH-109
300008-245EA 65.55 USD InStock
300008-245
Cylinder Holders, 4B's Bracket, ORS Nasco
Welding Supplies
  • The rubber gasket on the inside of the holder, eliminates all concern of friction and noise
  • Turns with your fingers tighten or loosen the wing nut, which holds the tank firmly in place, or lets the bolt swing out of your way
  • The yoke is welded to the cylinder holder and is fastened to the bracket with two (2) 3/4" long and 5/16" bolts, lock washers and nuts
  • The holder is hinged and swings back giving you access to the tank much quicker and easier than chains and makeshift methods
  • Strong enough to support connecting pipes and valves
  • Makes it easy to remove and replace
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty