Would you like to log in again?
Water Coolers, Anchor Brand
Supplier: Anchor Brand
100-R950V
300058-762EA 1488.59 USD InStock
300058-762
Water Coolers, Anchor Brand
Welding Supplies
  • 5/8" LH Std. inert gas fittings - 1/4" FPT
  • Rotary Gear Pump 1.7 gpm at 50 psi
  • Visible water level and water flow sight glass
  • Thermal overload
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty