Would you like to log in again?
Adjustable Wrench Sets, Proto®, ORS Nasco
Supplier: ORS Nasco
577-795
300003-166PK 172.85 USD InStock
300003-166
Adjustable Wrench Sets, Proto®, ORS Nasco
Wrenches
  • Large jaw capacity
  • Radius corner design increases strength and fastener surface contact area
  • Thin, slightly tapered jaws for access in tight work areas
  • Large serrated knurl for jaw adjustment
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty