Would you like to log in again?
Wire Strippers, Stanley®, ORS Nasco
Supplier: ORS Nasco
680-84-213
300033-674EA 99.7 USD InStock
300033-674
Wire Strippers, Stanley®, ORS Nasco
Cutters
Strippers to cut wires with a hole for bending and looping wire.
  • Adjustable slide stop and mechanical locking
  • Rust resistant finish
  • V Notches shaped jaw
For wire of 26 AWG (min) and 10 AWG (max).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty