Would you like to log in again?
Hand Drilling Hammers, Proto®, ORS Nasco
Supplier: ORS Nasco
577-1443G 577-1444G
300023-894EA 93.4 USD InStock
300023-894 300023-896
Hand Drilling Hammers, Proto®, ORS Nasco
Mallets and Hammers
For heavy blows in areas with limited swing arc on chisels, punches, star drills and hardened nails. Well balanced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty