Would you like to log in again?
Heat Guns, Milwaukee® Electric Tools
Supplier: Milwaukee® Electric Tools
495-8977-20 495-8975-6
300042-754EA 395.04 USD InStock
300042-754 300009-293
Heat Guns, Milwaukee® Electric Tools
Heaters Hot Air Dryers
Ideal for various applications from forming and molding plastic, thawing pipes, shrink fit plastic, and other applications that require a heat source.
  • Impact resistant heating element
  • Soft air velocity: Reaches required working surface temperature faster then conventional heat guns
  • Maintains temperature for fast completion of work
  • Cool air inlet design
  • Support stand
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty