Would you like to log in again?
Digital Multimeters, Milwaukee® Electric Tools
Supplier: Milwaukee Electric Tools
495-2217-20
300081-878EA 281.21 USD InStock
300081-878
Digital Multimeters, Milwaukee® Electric Tools
Electrical Meters Digital Multimeters
Simplified user interface feature set specifically for the electrician and HVAC technician.
  • Rugged overmolding for durability
  • Overmolded dial for one handed use
  • Slide rail for mounting voltage detector or magnet accessories
  • Functions include continuity, current, resistance, voltage, frequency
Ordering information: Includes instructions, test leads, multimeter, and thermocouple.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty