Would you like to log in again?
Electrician's Scissors, Klein Tools
Supplier: Klein Tools
409-2100-7
300002-390EA 50.1 USD InStock
300002-390
Electrician's Scissors, Klein Tools
Scissors
Designed for telecom and electrical applications. They cut wire, cable and cordage.
  • Upper blade has notches for stripping 19 AWG and 23 AWG wire
  • Strong, tempered steel
  • Nickel-plated finish resists corrosion
  • Scraper and file on outside of both blades
  • Serrated teeth allow for non-slip cutting action
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty