Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hand Carry-Electric Compressors, DeWalt®

Supplier: DeWalt
115-D55154
300060-408EA 915.43 USD InStock
300060-408
Hand Carry-Electric Compressors, DeWalt®
Pumps Compressors
Flow rate: 3.8 CFM @ 100 psi

  • Oil lubricated pump improves durability
  • Overload protection eliminates risk of motor damage
  • Dual soft start valves assist in cold weather start up
  • Cast iron cylinder enhances pump life
  • 2 pole induction motor
  • Amperes: 15 A

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty