Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Pear Head Ratchets, Proto®, ORS Nasco
Supplier: ORS Nasco
577-5449XL
300000-527EA 155.32 USD InStock
300000-527
Pear Head Ratchets, Proto®, ORS Nasco
Tool Sets
Sealed design helps prevents debris from entering ratchet mechanism reducing wear.
  • Low profile reversing lever design for quick, one hand operation
  • Knurled bands allow for increased grip and slip resistance
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty