Would you like to log in again?
Nalgene® Polypropylene Spigot Closure, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
6423-0010
27802-069CS 495.15 USD InStock
27802-069
Nalgene® Polypropylene Spigot Closure, Thermo Scientific
Bottles Carboys
Nalgene® polypropylene spigot closures can be used to replace spigots to provide a leakproof seal when autoclaving liquids in a carboy.
They are also useful when handling could damage or accidentally turn on spigot, or when spigot-less carboy is required. These spigot closures fit all Nalgene® carboys and selected Nalgene® tanks with spigots.
Ordering information: Included with all NALGENE® carboys with spigot.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty