Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG gamma Goat Polyclonal Antibody

Supplier: SeraCare
5210-0297
95058-920EA 337.44 USD InStock
95058-920
Anti-IgG gamma Goat Polyclonal Antibody
Antibodies
Secondary Goat Anti-IgG (g) F(ab')2 frag Reacts with Human

Type: Secondary
Antigen: IgG gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: gamma chain
Host: Goat
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty