Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-R. salmoninarum Antibody (FITC)

Supplier: SeraCare

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

5330-0063
95058-198EA 541.97 USD InStock
95058-198
Anti-R. salmoninarum Antibody (FITC)
Antibodies
Primary Goat Anti-R. salmoninarum Reacts with Renibacterium salmoninarum

Type: Primary
Antigen: R. salmoninarum
Clonality: Polyclonal
Clone:
Conjugation: Fluorescein (FITC)
Epitope:
Host: Goat
Isotype:
Reactivity: Renibacterium salmoninarum

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty