Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GFP Mouse Monoclonal Antibody (DyLight® 649) [clone: 9F9.F9]

Supplier: Rockland Immunochemicals
200-343-215 200-343-215
RL200-343-215EA 498 USD InStock
RL200-343-215 CA200-343-215
Anti-GFP Mouse Monoclonal Antibody (DyLight® 649) [clone: 9F9.F9]
Antibodies
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: ELISA: 1:20000-1:40000; FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000-1:25000;
Restore with deionized water (or equivalent); 100 µl

Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: DyLight® 649
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty