Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HA Rabbit Polyclonal Antibody (DyLight® 800)

Supplier: Rockland Immunochemicals
600-445-384 600-445-384
RL600-445-384EA 498 USD InStock
CA600-445-384 RL600-445-384
Anti-HA Rabbit Polyclonal Antibody (DyLight® 800)
Antibodies
Primary Rabbit Anti-HA EPITOPE TAG Reacts with Pan

Type: Tag
Antigen: HA
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pan

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty