Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-P53 Mouse Monoclonal Antibody

Supplier: EMD Millipore
OP33-20UG OP33-100UG OP33-20UG
EMOP33-20UGEA 160.75 USD InStock
CA80001-336 EMOP33-20UG CA80512-086
Anti-P53 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-p53 (Ab-5) (Wild type) (PAb1620) Reacts with cow (bovine, cattle), human, mouse, primate, rat.

Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Human, Mouse, Primate, Rat

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty