Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium chloride hexahydrate ≥99.999% (Al basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Aluminum chloride hexahydrate

13-1309-25G 13-1309-100G
100204-896EA 106.15 USD InStock
100204-896 100209-644
Aluminium chloride hexahydrate ≥99.999% (Al basis), PURATREM
Aluminium chloride hexahydrate


Formula: AlCl₃·6H₂O
MW: 241.43 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.398
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 1759
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty