Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylaluminium ≥93%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Triethylaluminum

13-1850-500G 13-1850-25G 13-1850-100G
100202-926EA 304.31 USD InStock
100202-926 100196-038 100203-730
Triethylaluminium ≥93%
Triethylaluminium
Metal Alkyls, Volatile Precursors for CVD


Formula: C₆H₁₅Al
MW: 114.17 g/mol
Boiling Pt: 186 °C
Melting Pt: –52.5 °C
Density: 0,835
Flash Pt: –1 °F
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3394
ADR: 4.2,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty