Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylphosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC)

15-4402-25G
100209-728EA 1593.72 USD InStock
100209-728
Phenylphosphine ≥99%
Phenylphosphine
Phosphine


Formula: C₆H₇P
Boiling Pt: 160 °C
Density: 1,001
Flash Pt: 165 °F
MDL Number: MFCD00002094
CAS Number: 638-21-1
EINECS: 211-325-4
UN: 2845
ADR: 4.2,I
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty