Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylphosphine ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Methyl phosphine

Sure/Seal™ bottle (Trademark of Sigma-Aldrich Co. LLC)

15-6502-25G
100198-140EA 702.03 USD InStock
100198-140
Trimethylphosphine ≥98%
Trimethylphosphine
Phosphine


Formula: C₃H₉P
MW: 76.08 g/mol
Boiling Pt: 38…40 °C
Melting Pt: –85 °C
Density: 0,748
Flash Pt: –22 °F
MDL Number: MFCD00008510
CAS Number: 594-09-2
EINECS: 209-823-1
UN: 1993
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty