Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium carbonate ≥99.997% (K basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Carbonic acid potassium saltCarbonic acid, dipotassium salt

19-2600-50G 19-2600-10G
100207-240EA 370.83 USD InStock
100207-240 100203-024
Potassium carbonate ≥99.997% (K basis), PURATREM
Potassium carbonate


Formula: K₂CO₃
MW: 138.21 g/mol
Boiling Pt: Dec.
Melting Pt: 891 °C
Density: 2.428
Storage Temperature: Ambient
MDL Number: MFCD00011382
CAS Number: 584-08-7
EINECS: 209-529-3
Merck Index: 12,07781
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/98/16640598.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty