Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylgermane ≥98%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Tetra-n-butylgermaniumTetra-n-butylgermane

32-2050-25G 32-2050-5G
100203-768EA 564.74 USD InStock
100203-768 100197-110
Tetrabutylgermane ≥98%
Tetrabutylgermane
Metal Alkyls, Volatile Precursors for CVD


Formula: C₁₆H₃₆Ge
MW: 301.07 g/mol
Boiling Pt: 130…133 °C (5 mmHg)
Density: 0,934
Storage Temperature: Ambient
MDL Number: MFCD00015224
CAS Number: 1067-42-1
EINECS: 213-929-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty