Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium ≥99.9%, dendritic pieces

Supplier: STREM CHEMICALS INC MS
Danger

38-0074-25G
100203-104EA 4084.8 USD InStock
100203-104
Strontium ≥99.9%, dendritic pieces
Strontium


Formula: Sr
Boiling Pt: 1384 °C
Melting Pt: 769 °C
Density: 2,54
MDL Number: MFCD00134060
CAS Number: 7440-24-6
EINECS: 231-133-4
UN: 1393
ADR: 4.3,II
Merck Index: 12,08994
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty