Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium lanthanum (III) oxide

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Lanthanum(III) aluminum oxideAluminium lanthanum trioxideLanthanum Aluminium oxide

57-0500-25G 57-0500-5G
100198-226EA 203.82 USD InStock
100198-226 100203-672
Aluminium lanthanum (III) oxide
Aluminium lanthanum (III) oxide


Formula: AlLaO₃
MW: 213.89 g/mol
MDL Number: MFCD00144550
CAS Number: 12003-65-5
EINECS: 234-433-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty