Would you like to log in again?
Corning® HYPERStack™ Stainless Steel Manipulator, Corning
Supplier: Corning
Corning® HYPERStack™
11000
89184-638CS 22925.28 USD InStock
89184-638
Corning® HYPERStack™ Stainless Steel Manipulator, Corning
Reaction Vessels Reaction Vessels, Flask Shape
The Corning® HYPERStack™ Stainless Steel Manipulator accommodates both HYPERStack™-36 and CellSTACK-10 vessels.
It handles two units of the same type simultaneously or one unit at a time. It is ergonomically designed and has large, easy to read dials that are clearly marked to identify positions for ease of use in protocol development.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty