Would you like to log in again?
Individual Protease Inhibitor Kit, G-Biosciences
Supplier: G-Biosciences
786-207 786-207
82022-640EA 477.83 USD InStock
CA82022-640 82022-640
Individual Protease Inhibitor Kit, G-Biosciences
Cell Culture Supplements and Additives
A selection of individual protease inhibitors are offered for researchers who wish to design their own cocktails, supplement existing cocktails, or screen for specific proteases
Also available is the Protease Inhibitor Set, which contains all 12 of our individual protease inhibitors, each in a ready-to-use solution at a 100X concentration. The 1X concentration of the protease inhibitors is designed to give >90% inhibition in crude tissue extracts. Various concentrations and/or combinations of protease inhibitors may be used to inhibit a broad spectrum of protease activity.
G-Biosciences offers the following individual protease inhibitors as a dry powder: AEBSF, ALLN, Antipain dihydrochloride, Aprotinin, Bestatin, Chymostatin, E-64, EDTA-Na2, Leupeptin, Pepstatin, Phosphoramidon, and PMSF.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty