Would you like to log in again?
Immobilized Pepsin
Supplier: G-Biosciences
786-791 786-791
89168-038EA 340.66 USD InStock
CA89168-038 89168-038
Immobilized Pepsin
Enzymes
Pepsin is a proteolytic enzyme that is routinely used for the generation of F(ab)2 fragments from immunoglobulin G (IgG)
The Immobilized Pepsin offers the distinct advantage of eliminating enzyme contamination of the F(ab)2 fragments. The F(ab)2 fragments can be purified from undigested IgG with Immobilized Protein A and can be further purified from the small Fc fragments by dialyze with a 50kDa MWCO membrane (Tube-O-DIALYZER™) or by gel filtration. Supplied as a 50% slurry in 50% glycerol, 0.1M sodium acetate, pH4.4 with sodium azide as a preservative.
Pepsin has the ability to cleave the heavy chains near the hinge region. One or more of the disulfide bonds that join the heavy chains in the hinge region are preserved, so the two Fab regions of the antibody remain joined together, yielding a divalent molecule (containing two antibody binding sites), hence the designation F(ab)2. The light chains remain intact and attached to the heavy chain, whereas the Fc fragment is digested into small peptides.
Ordering information: Accessories not included.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty