Would you like to log in again?
Corning® Octagonal PET Bottles, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Corning®
431734 431733 431732 431731 431730 431729
89204-720CS 189.19 USD InStock
89204-720 89235-040 89204-716 89204-718 89235-042 89204-714
Corning® Octagonal PET Bottles, Corning
Bottles Plastic Bottles
PET, clear, with natural HDPE screw cap.
  • Packaged in convenient, shrinkwrapped trays within an outer bag to assure cleanliness
  • Bottles are sterilized to 10⁻⁶ SAL and certified nonpyrogenic
  • USP class VI compliant bottles are validated noncytotoxic and nonhemolytic
ideal for storage and sampling of media, biologicals, and other aqueous solutions. The PET resin makes the bottle lightweight, break resistant and as transparent as glass. Leakproof, tamper-evident HDPE screw cap. Molded graduations for accurate measurements.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty