Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium stearate ≥95%

Supplier: STREM CHEMICALS INC MS
Synonyms: Stearic acid lithium saltLithium Stearate

93-0353-1000G 93-0353-250G
100199-468EA 285.91 USD InStock
100199-468 100200-018
Lithium stearate ≥95%
Lithium stearate


Formula: C₁₈H₃₅LiO₂
MW: 290.42 g/mol
Melting Pt: 220…221.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00042032
CAS Number: 4485-12-5
EINECS: 224-772-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty