Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lithium acetate dihydrate ≥98% (99.9%-Li)

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Acetic acid lithium salt

93-0362-1000G 93-0362-250G
100202-518EA 346.77 USD InStock
100202-518 100210-122
Lithium acetate dihydrate ≥98% (99.9%-Li)
Lithium acetate dihydrate


Formula: H₃CCOOLi·2H₂O
MW: 102.02 g/mol
Melting Pt: Dec.
Density: 1,3
Storage Temperature: Ambient
MDL Number: MFCD00066949
CAS Number: 6108-17-4
EINECS: 208-914-3
Merck Index: 13,05543
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty