Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitronium tetrafluoroborate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Nitryl tetrafluoroborate

93-0740-25G 93-0740-5G
100211-194EA 390.65 USD InStock
100211-194 100211-196
Nitronium tetrafluoroborate ≥97%
Nitronium tetrafluoroborate


Formula: BF₄NO₂
Melting Pt: 260 °C
Density: 1.26 g/cm³ (20 °C)
MDL Number: MFCD00011432
CAS Number: 13826-86-3
EINECS: 237-533-5
UN: 1759
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty