Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium nitrite ≥97% ACS

Supplier: 0000018203
Danger

93-1162-500G 93-1162-2000G
100201-860EA 309.97 USD InStock
100201-860 100201-862
Sodium nitrite ≥97% ACS
Sodium nitrite


Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320 °C (dec.)
Melting Pt: 271 °C
Density: 2.168
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 12,08793
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty