Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium hydroxide ≥95%

Supplier: STREM CHEMICALS INC MS

93-1229-500G 93-1229-2000G
100206-918EA 155.69 USD InStock
100206-918 100206-920
Magnesium hydroxide ≥95%
Magnesium hydroxide


Formula: Mg(OH)₂
MW: 58.32 g/mol
Density: 2.36
MDL Number: MFCD00011104
CAS Number: 1309-42-8
EINECS: 215-170-3
Merck Index: 13,05693
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty