Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(3-Aminopropyl)triethoxysilane ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: 3-(Triethoxysilyl)propylamineGeniosil GF 933-AminopropyltriethoxysilaneTriethoxy(3-aminopropyl)silane

93-1402-100G 93-1402-500G
100208-104EA 268.92 USD InStock
100208-104 100209-012
(3-Aminopropyl)triethoxysilane ≥98%
(3-Aminopropyl)triethoxysilane
Si, Silanes & Silanols for CVD & ALD, Volatile Precursors for CVD


Formula: H₂NCH₂CH₂CH₂Si(OCH₂CH₃)₃
MW: 221.37 g/mol
Boiling Pt: 217 °C
Melting Pt: < –70 °C
Density: 0,943
Flash Pt: 220 °F
Storage Temperature: Ambient
MDL Number: MFCD00008207
CAS Number: 919-30-2
EINECS: 213-048-4
UN: 1760
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty