Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyltrichlorosilane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Trichloro(methyl)silane

93-1417-100G 93-1417-500G
100197-702EA 43.88 USD InStock
100197-702 100205-332
Methyltrichlorosilane ≥97%
Methyltrichlorosilane


Formula: CH₃Cl₃Si
MW: 149.48 g/mol
Boiling Pt: 66.4 °C
Melting Pt: –77.8 °C
Density: 1.273 (25°)
Flash Pt: 5 °F
MDL Number: MFCD00000481
CAS Number: 75-79-6
EINECS: 200-902-6
UN: 1250
ADR: 3,II
REACH: 01-2119475226-36
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty