Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriethylsilane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Triethylchlorosilane

93-1463-25G 93-1463-5G
100201-918EA 72.18 USD InStock
100201-918 100206-008
Chlorotriethylsilane ≥99%
Chlorotriethylsilane


Formula: C₆H₁₅ClSi
Boiling Pt: 144 °C (735 mmHg)
Density: 0.8967 (20°)
Flash Pt: 85 °F
Storage Temperature: Ambient
MDL Number: MFCD00000507
CAS Number: 994-30-9
EINECS: 213-615-6
UN: 2920
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty