Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotriphenylsilane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Triphenylchlorosilane

93-1466-10G 93-1466-50G
100197-706EA 67.94 USD InStock
100197-706 100209-016
Chlorotriphenylsilane ≥97%
Chlorotriphenylsilane


Formula: C₁₈H₁₅ClSi
MW: 294.86 g/mol
Boiling Pt: 378 °C
Melting Pt: 91…92 °C
Density: 1.0165 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000496
CAS Number: 76-86-8
EINECS: 200-989-0
UN: 1759
ADR: 8,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty