Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus(III) chloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Phosphorus trichloride

93-1587-250G
100201-940EA 103.32 USD InStock
100201-940
Phosphorus(III) chloride ≥98%
Phosphorus (III) chloride
Halides for CVD & ALD


Formula: Cl₃P
MW: 137.33 g/mol
Boiling Pt: 76 °C
Melting Pt: –111.8 °C
Density: 1.574
Storage Temperature: Ambient
MDL Number: MFCD00011438
CAS Number: 7719-12-2
EINECS: 231-749-3
UN: 1809
ADR: 6.1,I
Merck Index: 12,07515
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty