Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iron(III) sulfate hydrate, Technical Grade

Supplier: 0000018203
Warning
Synonyms: Ferric sulfate hydrateDiiron trissulfate hydrateDiiron trissulphate hydrateFerric sulphate hydrate

93-2623-1000G 93-2623-250G
100207-812EA 59.45 USD InStock
100207-812 100197-772
Iron(III) sulfate hydrate, Technical Grade
Iron (III) sulphate hydrate


Formula: Fe₂(SO₄)₃·xH₂O
MW: 399.88 g/mol (anhy)
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149714
CAS Number: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty