Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Titanium(IV) oxide anatase, nanopowder

Supplier: STREM CHEMICALS INC MS

22-1400-25G 22-1400-5G
101433-190EA 60.86 USD InStock
101433-190 101433-188
Titanium(IV) oxide anatase, nanopowder
Titanium (IV) oxide anatase
Nanomaterials


Formula: TiO₂
MW: 79.88 g/mol
Boiling Pt: 2900 °C (1013 hPa)
Melting Pt: 1855 °C
Density: 4.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011269
CAS Number: 1317-70-0
EINECS: 236-675-5
Merck Index: 12,09612
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty