Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichlorodiethylgermane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: DichlorodiethylgermaneDiethylgermanium dichloride

93-3220-1G 93-3220-5G
100202-090EA 376.49 USD InStock
100202-090 100210-364
Dichlorodiethylgermane ≥97%
Dichlorodiethylgermane


Formula: C₄H₁₀Cl₂Ge
Boiling Pt: 175 °C
Density: 1.372
MDL Number: MFCD00013589
CAS Number: 13314-52-8
EINECS: 236-344-5
UN: 1760
ADR: 8,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty