Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylgermane ≥99%

Supplier: STREM CHEMICALS INC MS
Danger
Synonyms: Tetraethylgermanium

93-3227-2G 93-3227-10G
100208-158EA 509.54 USD InStock
100208-158 100207-038
Tetraethylgermane ≥99%
Tetraethylgermane
Metal Alkyls, Volatile Precursors for CVD


Formula: C₈H₂₀Ge
Boiling Pt: 165.5 °C
Melting Pt: –90 °C
Density: 1.1989
Flash Pt: 85 °F
MDL Number: MFCD00015096
CAS Number: 597-63-7
EINECS: 209-905-7
UN: 1993
ADR: 3,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty