Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rubidium acetate ≥99% (Rb basis)

Supplier: STREM CHEMICALS INC MS

93-3701-50G 93-3701-10G
100209-098EA 588.8 USD InStock
100209-098 100202-104
Rubidium acetate ≥99% (Rb basis)
Rubidium acetate


Formula: H₃CCOORb
MW: 144.51 g/mol
Melting Pt: 246 °C
MDL Number: MFCD00013058
CAS Number: 563-67-7
EINECS: 209-255-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty