Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium carbonate ≥97% (contains ~1.0-2.5% barium carbonate)

Supplier: STREM CHEMICALS INC MS

93-3804-1000G 93-3804-250G
100196-636EA 106.15 USD InStock
100196-636 100204-318
Strontium carbonate ≥97% (contains ~1.0-2.5% barium carbonate)
Strontium carbonate


Formula: CO₃Sr
MW: 147.63 g/mol
Boiling Pt: 2647 °C (1 mmHg)
Melting Pt: 1497 °C
Density: 3.70
Storage Temperature: Ambient
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7
Merck Index: 14,08838
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty