Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum(VI) oxide ≥99.5%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Molybdenum trioxide

93-4215-250G 93-4215-50G
100197-858EA 38.22 USD InStock
100197-858 100207-060
Molybdenum(VI) oxide ≥99.5%
Molybdenum (VI) oxide


Formula: MoO₃
MW: 143.94 g/mol
Boiling Pt: (subl.)
Melting Pt: 795 °C
Density: 4.69
MDL Number: MFCD00003469
CAS Number: 1313-27-5
EINECS: 215-204-7
UN: 3288
ADR: 6.1,III
Merck Index: 12,06321
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty