Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium tetrabromopalladate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning
Synonyms: Potassium tetrabromopalladate (II)Potassium tetrabromopalladate(II)

93-4611-5G 93-4611-1G
100197-862EA 508.12 USD InStock
100197-862 100197-860
Dipotassium tetrabromopalladate ≥98%
Dipotassium tetrabromopalladate


Formula: Br₄K₂Pd
MW: 504.23 g/mol
MDL Number: MFCD00049659
CAS Number: 13826-93-2
EINECS: 237-535-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty