Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver bromide ≥99.9% (Ag basis)

Supplier: STREM CHEMICALS INC MS
Synonyms: Argentous bromide

93-4705-50G 93-4705-10G
100207-080EA 101.91 USD InStock
100207-080 100200-148
Silver bromide ≥99.9% (Ag basis)
Silver bromide


Formula: AgBr
MW: 187.77 g/mol
Boiling Pt: 1300 °C (1013 hPa)
Melting Pt: 432 °C
Density: 6.473
Storage Temperature: Ambient
MDL Number: MFCD00003398
CAS Number: 7785-23-1
EINECS: 232-076-8
Merck Index: 12,08649
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty