Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver(I) thiocyanate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

93-4735-25G 93-4735-5G
100198-384EA 418.95 USD InStock
100198-384 100196-682
Silver(I) thiocyanate ≥99%
Silver (I) thiocyanate


Formula: AgSCN
MW: 165.95 g/mol
Melting Pt: Dec.
MDL Number: MFCD00003416
CAS Number: 1701-93-5
EINECS: 216-934-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty