Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyldiphenyltin

Supplier: STREM CHEMICALS INC MS
Warning

93-5015-5G 93-5015-25G
100204-354EA 175.51 USD InStock
100204-354 100202-174
Dimethyldiphenyltin
Dimethyldiphenyltin


Formula: C₁₄H₁₆Sn
MDL Number: MFCD00058812
CAS Number: 1080-43-9
UN: 2788
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty